Anti-CD7 Rabbit Polyclonal Antibody
Catalog # BSBTA01974-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:CD7
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for CD7 detection. Tested with IHC-P, FCM, Direct ELISA in Human.
Type: Primary
Antigen: CD7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human