Anti-IGFBP5 Rabbit Polyclonal Antibody
Catalog # BSBTA01952-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Insulin-like growth factor-binding protein 5
- Antigen symbol:IGFBP5
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Insulin-like growth factor-binding protein 5(IGFBP5) detection. Tested with WB, ELISA in Human.
Type: Primary
Antigen: IGFBP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human