Anti-SPARC Rabbit Polyclonal Antibody
BSBTA00862-1
- Antibody type:Primary
- Antigen symbol:SPARC
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for SPARC(SPARC) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat