Anti-IGFBP1 Rabbit Polyclonal Antibody
BSBTA00922
- Antibody type:Primary
- Antigen name:Insulin-like growth factor-binding protein 1
- Antigen symbol:IGFBP1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Insulin-like growth factor-binding protein 1(IGFBP1) detection. Tested with WB, IHC-P in Human.
Type: Primary
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human