Peel-A-Way® Embedding Moulds
Supplier: POLYSCIENCES
Certificates
About this item
These disposable paraffin moulds are designed to 'Peel-A-Way' from the solidified embedded block.
- Versatile and can be used with polyfreeze, JB-4, GMA, MMA and epoxy
- Allows the specimen to freeze to the chuck without defrosting the base
- Unique polyethylene construction eliminates the need for release agents and block trimming
Useful for frozen tissue where defrosting during transfer to cryostat is an issue.
Frequently Bought Together