Anti-FSH-R Rabbit Polyclonal Antibody
Catalog # BSBTA00897
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Follicle-stimulating hormone receptor
- Antigen symbol:FSH-R
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Follicle-stimulating hormone receptor(FSH-R)(FSHR) detection. Tested with WB in Human;Mouse.
Type: Primary
Antigen: FSH-R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse