Anti-DVL2 Rabbit Polyclonal Antibody
BSBTA02404-3
- Antibody type:Primary
- Antigen name:Segment polarity protein dishevelled homolog DVL-2
- Antigen symbol:DVL2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Segment polarity protein dishevelled homolog DVL-2(DVL2) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: DVL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat