Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test sieves, 203×50 mm
Supplier:   Retsch GmbH
Test sieves, 203×50 mm
Supplier:  Retsch GmbH
Certificates
Test sieves, 203×50 mm
42 Options Available Below
Product Details & Documents
Sieves made from 304 stainless steel.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieves are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.
 : ISO 3310-1:2001-09
Product Family Options
Product Information
  • Ø
    Height
    Mesh size
    Availability
    Price
  • 203 mm
    50 mm
    0,020 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.260,00 kr
    9.260,00 kr
    1 * 1 Items
       9.260,00 kr
  • 203 mm
    50 mm
    0,025 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.130,00 kr
    5.130,00 kr
    1 * 1 Items
       5.130,00 kr
  • 203 mm
    50 mm
    0,032 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.200,00 kr
    3.200,00 kr
    1 * 1 Items
       3.200,00 kr
  • 203 mm
    50 mm
    0,036 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.200,00 kr
    3.200,00 kr
    1 * 1 Items
       3.200,00 kr
  • 203 mm
    50 mm
    0,038 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.200,00 kr
    3.200,00 kr
    1 * 1 Items
       3.200,00 kr
  • 203 mm
    50 mm
    0,040 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.430,00 kr
    2.430,00 kr
    1 * 1 Items
       2.430,00 kr
  • 203 mm
    50 mm
    0,045 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.430,00 kr
    2.430,00 kr
    1 * 1 Items
       2.430,00 kr
  • 203 mm
    50 mm
    0,050 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.120,00 kr
    2.120,00 kr
    1 * 1 Items
       2.120,00 kr
  • 203 mm
    50 mm
    0,053 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.120,00 kr
    2.120,00 kr
    1 * 1 Items
       2.120,00 kr
  • 203 mm
    50 mm
    0,056 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.950,00 kr
    1.950,00 kr
    1 * 1 Items
       1.950,00 kr
  • 203 mm
    50 mm
    0,063 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,071 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,075 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,080 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,090 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,100 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,106 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,112 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,140 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,160 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,180 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,200 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,212 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,224 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,280 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,315 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,355 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,400 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,425 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,450 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,500 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.790,00 kr
    1.790,00 kr
    1 * 1 Items
       1.790,00 kr
  • 203 mm
    50 mm
    0,560 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,600 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,630 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,710 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,800 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,850 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr
  • 203 mm
    50 mm
    0,900 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.820,00 kr
    1.820,00 kr
    1 * 1 Items
       1.820,00 kr