Order Entry
Denmark
ContactUsLinkComponent
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, Dr. Ehrenstorfer
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, Dr. Ehrenstorfer
Catalog # EHERC15089000
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2-Methyl-4-isothiazolin-3-one, Dr. Ehrenstorfer
Catalog # EHERC15089000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Standards:
    Reference Standard
  • CAS No.:
    2682-20-4
  • Description:
    2-Methyl-4-isothiazolin-3-one
  • Formula:
    C₄H₅NOS
  • Description:
    2-Methyl-4-isothiazolin-3-one

Specifications

About this item