Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for flame atomisation systems
Accessories for flame atomisation systems
Supplier:   VARIAN
Accessories for flame atomisation systems
Supplier:  VARIAN
Certificates
Accessories for flame atomisation systems
Accessories for flame atomisation systems
8 Options Available Below
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Product Family Options
Product Information
  • Availability
    Price
  • VARI9910093000
    Nebuliser capillary kit
    Nebuliser capillary kit
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.550,00 kr
    7.550,00 kr
    1 * 1 Items
       7.550,00 kr
  • VARI9910025700
    Glass impact beads
    Glass impact beads
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 2.010,00 kr
    2.010,00 kr
    1 * 5 Items
       2.010,00 kr
  • VARI9910093600
    Fluorinated mixing paddles
    Fluorinated mixing paddles
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.480,00 kr
    2.480,00 kr
    1 * 1 Items
       2.480,00 kr
  • VARI9910093400
    Aqueous spray chamber o-ring
    Aqueous spray chamber o-ring
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 846,00 kr
    846,00 kr
    1 * 1 Items
       846,00 kr
  • VARI9910093500
    Organics o-ring
    Organics o-ring
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11.490,00 kr
    11.490,00 kr
    1 * 1 Items
       11.490,00 kr
  • VARI6910009700
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • VARI9910093100
    Integrated nebuliser block
    Integrated nebuliser block
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 13.360,00 kr
    13.360,00 kr
    1 * 1 Items
       13.360,00 kr
  • VARI9910105800
    Integrated liquid trap (fluorinated float and magnet)
    Integrated liquid trap (fluorinated float and magnet)
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 4.190,00 kr
    4.190,00 kr
    1 * 1 Items
       4.190,00 kr