Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standards, vardenafils, Ph. Eur., RS
 :  EDQM
 
Organic reference standards, vardenafils, Ph. Eur., RS
 :  EDQM

 

  • CAS No.:
    330808-88-3
  • Formula:
    C₂₃H₃₉ClN₆O₇S
  • Description:
    Vardenafil hydrochloride

 

Frequently Bought Together