Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, tretinoins, Ph. Eur., RS
Organic reference standard, tretinoins, Ph. Eur., RS
Organic reference standard, tretinoins, Ph. Eur., RS
Catalog # EDQMT1850000
Supplier:
EDQM
CAS Number:
Expand image
Organic reference standard, tretinoins, Ph. Eur., RS
Catalog # EDQMT1850000
Supplier:
EDQM
CAS Number:
Specifications
Pk:
50 mg
Standards:
Reference standard
CAS No.:
302-79-4
Description:
Tretinoin
Formula:
C₂₀H₂₈O₂
Description:
Tretinoin
Specifications
About this item
Product Details
Product Details