Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, thiamazole, Ph. Eur., RS
Organic reference standards, thiamazole, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, thiamazole, Ph. Eur., RS
Supplier:  EDQM
Certificates

Specifications

  • CAS No.:
    60-56-0
  • Formula:
    C₄H₆N₂S
  • Description:
    Thiamazole

Specifications

Related Information

Frequently Bought Together