Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, sodium alendronates, Ph. Eur., RS
Organic reference standards, sodium alendronates, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, sodium alendronates, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    121268-17-5
  • Formula:
    C₄H₁₈NNaO₁₀P₂
  • Description:
    Sodium alendronate trihydrate

Specifications

Frequently Bought Together