Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, ranitidine, Ph. Eur., RS
Organic reference standards, ranitidine, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, ranitidine, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    66357-59-3
  • Formula:
    C₁₃H₂₃ClN₄O₃S
  • Description:
    Ranitidine for impurity A identification

Specifications

Frequently Bought Together