Organic reference standard, Racemic methyldopa, Ph. Eur., RS
Catalog # EDQMY0001374
Supplier: EDQM
CAS Number:
Specifications
- Pk:5 mg
- Standards:Reference standard
- CAS No.:41372-08-1
- Description:Racemic methyldopa
- Formula:C₁₀H₁₅NO₅
- Description:Racemic methyldopa