Organic reference standards, potassium clavulanate, Ph. Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:374816-32-7
- Formula:C₁₃H₁₅NO₆
- Description:Potassium clavulanate impurity G, 4-[[(1S)-1-carboxy-2-(4-hydroxyphenyl)ethyl]amino]-4-oxobutanoic acid (N-(hydrogensuccinyl)tyrosine)
Specifications
Frequently Bought Together