Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, oseltamivir, Ph. Eur., RS
Organic reference standards, oseltamivir, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, oseltamivir, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    814-29-9
  • Formula:
    C₁₂H₂₇OP
  • Description:
    Oseltamivir impurity H, tributylphosphane oxide

Specifications

Frequently Bought Together