Organic reference standards, olmesartan medoxomil, Ph. Eur. RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:144689-63-4
- Formula:C₂₉H₃₀N₆O₆
- Description:Olmesartan medoxomil for system suitability
Specifications
Frequently Bought Together