Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, glucosamine, Ph. Eur. RS
Organic reference standards, glucosamine, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, glucosamine, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    38899-05-7
  • Formula:
    C₆H₁₂NNaO₈S
  • Description:
    Glucosamine sulfate sodium chloride

Specifications

Frequently Bought Together