Organic reference standards, desloratadine, Ph. Eur. RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:100643-71-8
- Formula:C₁₉H₁₉ClN₂
- Description:Desloratadine for system suitability
Specifications
Frequently Bought Together