Order Entry
Denmark
ContactUsLinkComponent
ASB-C8Ø (4-n-Octylbenzoylamidopropyldimethylammonio sulphobetaine)
ASB-C8Ø (4-n-Octylbenzoylamidopropyldimethylammonio sulphobetaine)
Catalog # DG064
Supplier:  G-Biosciences
CAS Number:  
ASB-C8Ø (4-n-Octylbenzoylamidopropyldimethylammonio sulphobetaine)
Catalog # DG064
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :440.6 g/mol
CAS Number:216667-49-1
Formula:C₂₃H₄₀N₂O₄S