Order Entry
Denmark
Orders LinkContactUsLinkComponent
ASB-14 ≥99%

ASB-14 ≥99%

5 g
Catalog # DG061
Supplier:   G-Biosciences
ASB-14 ≥99%
Catalog # DG061
Supplier:  G-Biosciences
CAS Number:  216667-08-2
ASB-14 ≥99%

5 g

CAS Number:  216667-08-2
Price
4.120,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Zwitterionic detergent, Aminosulfobetaine with C14 alkyl tail is useful for solubilizing proteins for 2D analysis. Optimal solubility achieved in urea-thiourea mixtures and not in urea alone.

Reported to show better protein solubilization properties than CHAPS. ASB-14 has been shown to solubilise membrane proteins previously undetected.

Synonyms:  Amidosulphobetaine-14
  • CAS Number:216667-08-2
  • Formula:C₂₂H₄₆N₂O₄S
  • Molecular Weight:434.68 g/mol
Specifications

Specifications

  • Pk:
    5 g

Specifications