Organic reference standards, aciclovir, Ph. Eur. RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:59277-89-3
- Formula:C₈H₁₁N₅O₃
- Description:Aciclovir for peak identification 2
Specifications
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
2,3,4,6-Tetrachlorophenol analytical standard, Supelco®
Supplier: Merck
Conductivity standards
Supplier: Hach
n-Heptane, Supelco®
Supplier: Merck
IBUPROFEN FOR PEAK IDENTIFICATION 1 * 0,09 mg
Supplier: EDQM
VWR®, COD calibration standards, Certified Reference Materials (CRMs)
Supplier: VWR Chemicals
Fluquinconazole certified 1 * 0,1 g
Supplier: EHRENSTORFER
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
Supplier: Thermo Fisher Scientific
Penflufen Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
Multi-element standard solution 6 for ICP, TraceCERT®
Supplier: Merck