Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standards, Oleic Acid, Ph. Eur. RS
Organic reference standards, Oleic Acid, Ph. Eur. RS
Organic reference standards, Oleic Acid, Ph. Eur. RS
Catalog # EDQMY0001479
Supplier:
EDQM
CAS Number:
Expand image
Organic reference standards, Oleic Acid, Ph. Eur. RS
Catalog # EDQMY0001479
Supplier:
EDQM
CAS Number:
Specifications
Pk:
2 mg
Standards:
Reference standard
CAS No.:
112-80-1
Description:
Oleic acid
Formula:
C₁₈H₃₄O₂
Description:
Oleic acid
Specifications
About this item
Product Details
Product Details