Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, levonorgestrels, Ph. Eur.
Organic reference standards, levonorgestrels, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, levonorgestrels, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    797-63-7
  • Formula:
    C₂₁H₂₈O₂
  • Description:
    Levonorgestrel

Specifications

Frequently Bought Together