Organic reference standards, levetiracetams, Ph. Eur.
: EDQM
- CAS No.:102767-28-2
- Formula:C₈H₁₄N₂O₂
- Description:Levetiracetam
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Dimethyl disulphide
: EHRENSTORFER
1-Octene, Supelco®
: Merck
1H,1H,2H,2H-Perfluoro-1-octanol 1 * 100 mg
: EHRENSTORFER
VWR®, Flow Cup viscosity standards
: VWR Chemicals
Dichlobenil
: EHRENSTORFER
VWR®, COD calibration standards
: VWR Chemicals
Estradiol
: EHRENSTORFER
Buffer solutions
: REAGECON
Nitrite, (CRM), Supelco®
: Merck