VWR®, Glass Stoppers for Standard Ground Joints
Supplier: VWR Collection
Certificates
About this item
Borosilicate 3.3 glass.
- Hollow, with hexagonal grip
- Trip-tip to ensure every last drop of liquid returns into the vessel
- Resistant to chemicals and thermal shock
Certifications: DIN 12252
Frequently Bought Together