Ordreindtastning
Denmark
ContactUsLinkComponent
Organic reference standards, imatinibs, Ph. Eur.
Organic reference standards, imatinibs, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, imatinibs, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS-nr:
    350-03-8
  • Formel:
    C₇H₇NO
  • Beskrivelse:
    Imatinib impurity H, 1-(pyridin-3-yl)ethan-1-one

Specifications

Frequently Bought Together