Anti-IgG Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 35-8078-M002
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Rabbit
- Epitope:Heavy and light chain
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Specifications
About this item
Sheep anti-Rabbit IgG (H&L) - Affinity Pure, FITC Conjugate
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Sheep
Isotype:
Reactivity: Rabbit