Order Entry
Denmark
ContactUsLinkComponent
Test kits, photometric, Chlorine, free, LCK cuvette tests
Test kits, photometric, Chlorine, free, LCK cuvette tests
Catalog # HACHLCK410
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, Chlorine, free, LCK cuvette tests
Catalog # HACHLCK410
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    DPD
  • Pk:
    24 Tests
  • Description:
    Chlorine, free
  • Seq:
    212
  • Type:
    LCK410
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,05 - 2,0 mg/l Chlorine free/ClO₂
Specifications
About this item
Cuvette test for the determination or free chlorine, 0,05 to 2,0 mg/L Cl₂ and chlorine dioxide, 0,09 to 3,8 mg/L ClO₂. Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Certifications: ISO 7393-1-2-1985, DIN 38408 G4-2