Order Entry
Denmark
ContactUsLinkComponent
Alkaline phosphatase assay kit
Alkaline phosphatase assay kit
Catalog # ABNOKA1642
Supplier:  Abnova
CAS Number:  
undefined
Alkaline phosphatase assay kit
Catalog # ABNOKA1642
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Alkaline Phosphatase Assay Kit
  • Tests per kit:
    250

Specifications

About this item

Alkaline phosphatase assay kit allows the colorimetric kinetic determination of serum alkaline phosphatase activity.

  • High sensitivity and wide linear range: Use 5 µl serum or plasma sample; detection limit is 2 U/L, linear up to 800 U/L
  • Homogeneous and simple procedure: Simple 'mix-and-measure' procedure allows reliable quantitation of ALP activity within 5 minutes
  • Robust and amenable to HTS: All reagents are compatible with high-throughput liquid handling instruments

Applications:
Direct Assays: ALP activity in serum, plasma and other sources
Characterisation and quality control for ALP production
Drug Discovery: High throughput screening for ALP inhibitors and evaluation of ALP inhibitors

Delivery: Kit contains assay buffer 50 ml (pH 10,5), Mg Acetate 1,5 ml (0,2 M), pNPP liquid 600 µl (1 M) and Tartrazine calibrator10 ml.