Order Entry
Denmark
ContactUsLinkComponent
 
Sodium acetate, anhydrous ≥99.0%, white powder, AR® ACS, Macron Fine Chemicals™
 
undefined
Sodium acetate, anhydrous ≥99.0%, white powder, AR® ACS, Macron Fine Chemicals™
 

 

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (CH₃COONa):
    ≥99.0 %
  • ACS - pH of 5% Solution at 25°C:
    7.0 - 9.2
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Loss on Drying at 120°C:
    ≤1.0 %
  • ACS - Chloride (Cl):
    ≤0.002 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • Appearance (fine, white powder):

 

 

Frequently Bought Together