Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Hydrochloric acid 36,5 - 38,0%, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
 
 
Hydrochloric acid 36,5 - 38,0%, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Store below 25°C:
    Passes Test
  • Appearance:
    Passes Test
  • Assay (as HCl) (by acid-base titrn):
    36.5 - 38.0 %
  • Color (APHA):
    ≤ 10
  • Extractable Organic Substances:
    ≤ 5 ppm
  • Free Chlorine (as Cl₂):
    ≤ 1 ppm
  • Residue after Ignition:
    ≤ 3 ppm
  • Specific Gravity at 60°/60°F:
    1.185 - 1.192
  • Bromide (Br):
    ≤ 0.005 %
  • Phosphate (PO₄):
    ≤ 1 ppm
  • Sulfate (SO₄):
    ≤ 0.5 ppm
  • Sulfite (SO₃):
    ≤ 0.8 ppm
  • Ammonium (NH₄):
    ≤ 3 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 100.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 5.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 50 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 200.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 100.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 100.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 100.0 ppb
  • Heavy Metals (as Pb):
    ≤ 100 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 100.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 50 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 300.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 300.0 ppb
  • Trace Impurities - Mercury (Hg):
    ≤ 5.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 100.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 300.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 300 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 300.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 300.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 100.0 ppb

 

Frequently Bought Together