Organic reference standards, fluorouracils, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:56177-80-1
- Formula:C₆H₇FN₂O₂
- Description:Fluorouracil impurity F, 2-ethoxy-5-fluoropyrimidin-4(1H)-one (2-ethoxy-5-fluorouracil)
Specifications
Frequently Bought Together