Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, enrofloxacin, Ph. Eur.
Organic reference standards, enrofloxacin, Ph. Eur.
Catalog # EDQMY0001259
Supplier:  EDQM
CAS Number:  
Organic reference standards, enrofloxacin, Ph. Eur.
Catalog # EDQMY0001259
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Reference standard
  • CAS No.:
    93106-60-6
  • Description:
    Enrofloxacin, Ph. Eur. standard
  • Formula:
    C₁₉H₂₂FN₃O₃
  • Description:
    Enrofloxacin

Specifications

About this item