Organic reference standards, dexchlorpheniramines, Ph. Eur.
Catalog # EDQMD0720800
Supplier: EDQM
CAS Number:
Specifications
- Pk:60 mg
- Standards:Reference standard
- CAS No.:2438-32-6
- Description:Dexchlorpheniramine maleate, Ph. Eur. standard
- Formula:C₂₀H₂₃ClN₂O₄
- Description:Dexchlorpheniramine maleate