Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, desoxycortone acetate, Ph. Eur.
Organic reference standards, desoxycortone acetate, Ph. Eur.
Catalog # EDQMD0400000
Supplier:  EDQM
CAS Number:  
Organic reference standards, desoxycortone acetate, Ph. Eur.
Catalog # EDQMD0400000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    56-47-3
  • Description:
    Desoxycortone acetate, Ph. Eur. standard
  • Formula:
    C₂₃H₃₂O₄
  • Description:
    Desoxycortone acetate

Specifications

About this item