Order Entry
Denmark
Orders LinkContactUsLinkComponent
Organic reference standards, clindamycin phosphate, Ph. Eur.

Organic reference standards, clindamycin phosphate, Ph. Eur.

Clindamycin phosphate for system suitability, Ph. Eur. standard, Reference standard, 10 mg
Catalog # EDQMY0001680
Supplier:   EDQM
Organic reference standards, clindamycin phosphate, Ph. Eur.
Catalog # EDQMY0001680
Supplier:  EDQM
CAS Number:  
Organic reference standards, clindamycin phosphate, Ph. Eur.

Clindamycin phosphate for system suitability, Ph. Eur. standard, Reference standard, 10 mg

CAS Number:  
Price
1.740,00 kr
Log in for availability
1 * 10 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • Description:
    Clindamycin phosphate for system suitability, Ph. Eur. standard
  • Cat. No.:
    EDQMY0001680
  • Formula:
    C₁₈H₃₄ClN₂O₈PS
  • Description:
    Clindamycin phosphate for system suitability
  • CAS No.:
    24729-96-2

Specifications