Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, benzbromarone, Ph. Eur.
Organic reference standards, benzbromarone, Ph. Eur.
Catalog # EDQMY0000775
Supplier:  EDQM
CAS Number:  
Organic reference standards, benzbromarone, Ph. Eur.
Catalog # EDQMY0000775
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    3562-84-3
  • Description:
    Benzbromarone, Ph. Eur. standard
  • Formula:
    C₁₇H₁₂Br₂O₃
  • Description:
    Benzbromarone

Specifications

About this item