Organic reference standards, dorzolamides, Ph. Eur.
: EDQM
- CAS No.:120279-95-0
- Formula:C₁₀H₁₆N₂O₄S₃
- Description:Dorzolamide impurity A, (4R,6R)-4-(ethylamino)-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulphonamide-7,7-dioxide, Ph. Eur. standard
Frequently Bought Together