Order Entry
Denmark
ContactUsLinkComponent
 
Organic reference standards, docetaxels, Ph. Eur.
 :  EDQM
 
Organic reference standards, docetaxels, Ph. Eur.
 :  EDQM

 

  • CAS No.:
    32981-86-5
  • Formula:
    C₂₉H₃₆O₁₀
  • Description:
    Docetaxel impurity E, 5ß,20-epoxy-4-(acetyloxy)-1,7ß,10ß,13α-tetrahydroxy-9-oxotax-11-en-2α-yl benzoate (10-desacetyl-baccatin III), Ph. Eur. standard

 

Frequently Bought Together