Order Entry
Denmark
ContactUsLinkComponent
 
Pocket pH meters, LAQUAtwin pH-11 / pH-22 / pH-33
 :  HORIBA
 
undefined
Pocket pH meters, LAQUAtwin pH-11 / pH-22 / pH-33
 :  HORIBA

 

  • Catalog No:
  • 662-2292
  • 662-2290
  • 662-2291
  • Model:
  • pH-33
  • pH-11
  • pH-22
  • pH range:
  • 0,00 - 14,00
  • 0,0 - 14,0
  • 0,00 - 14,00
  • pH resolution:
  • 0,01
  • 0,1
  • 0,01
  • pH accuracy:
  • ± 0,01
  • ± 0,1
  • ± 0,01
  • pH calibration:
  • 5-point
  • 2-point
  • 3-point
  • Automatic buffer recognition:
  • USA: 1,68 / 4,01 / 7,00 / 10,01 / 12,46
    NIST: 1,68 / 4,01 / 6,86 / 9,18 / 12,46
  • USA: 1,68 / 4,01 / 7,00 / 10,01 / 12,46
    NIST: 1,68 / 4,01 / 6,86 / 9,18 / 12,46
  • USA: 1,68 / 4,01 / 7,00 / 10,01 / 12,46
    NIST: 1,68 / 4,01 / 6,86 / 9,18 / 12,46
  • mV range:
  • ± 650
  • ± 650
  • ± 650
  • mV resolution:
  • 1
  • 1
  • 1
  • Temperature range:
  • 0...+50,0 °C
  • -
  • -
  • Temperature resolution:
  • 0,1 °C
  • - °C
  • - °C
  • Display:
  • Custom (monochrome) digital LCD with backlight
  • Custom (monochrome) digital LCD with backlight
  • Custom (monochrome) digital LCD with backlight
  • Power supply:
  • 2× CR2032 batteries
  • 2× CR2032 batteries
  • 2× CR2032 batteries
  • Compliance:
  • IP67
  • IP67
  • IP67
  • W×D×H:
  • 164×29×20 mm
  • 164×29×20 mm
  • 164×29×20 mm
  • Weight:
  • 0,055 kg
  • 0,055 kg
  • 0,055 kg

 

Frequently Bought Together