Organic reference standards, azathioprines, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:4531-54-8
- Formula:C₄H₆N₄O₁
- Description:Azathioprine impurity A, 1-methyl-4-nitro-1H-imidazol-5-amine
Specifications
Frequently Bought Together