Order Entry
Denmark
ContactUsLinkComponent
Organic reference standards, alprenolol hydrochloride, Ph. Eur.
Organic reference standards, alprenolol hydrochloride, Ph. Eur.
Catalog # EDQMA0360000
Supplier:  EDQM
CAS Number:  
Organic reference standards, alprenolol hydrochloride, Ph. Eur.
Catalog # EDQMA0360000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    13707-88-5
  • Description:
    Alprenolol hydrochloride, Ph. Eur. standard
  • Formula:
    C₁₅H₂₄ClNO₂
  • Description:
    Alprenolol hydrochloride

Specifications

About this item