Order Entry
Denmark
ContactUsLinkComponent
Benzyl alcohol ≥99.5% (by GC), SupraSolv® for headspace gas chromatography
Benzyl alcohol ≥99.5% (by GC), SupraSolv® for headspace gas chromatography
Supplier:  Merck
Certificates
undefined
Benzyl alcohol ≥99.5% (by GC), SupraSolv® for headspace gas chromatography
Supplier:  Merck
Certificates

Specification Test Results

  • Purity (GC):
    ≥99,5%
  • Identity (IR):
    Conforms
  • Color:
    ≤10 Hazen
  • Acidity:
    ≤0,001 mEq/g
  • Alkalinity:
    ≤0,002 mEq/g
  • Water:
    ≤0,1%
  • GC/HS (Every residual solvent of class 1 acc. ICH):
    ≤1 µg/g
  • GC/HS (Every residual solvent of class 2 acc. ICH):
    ≤10 µg/g
  • GC/HS (Every residual solvent of class 3 acc. ICH):
    ≤50 µg/g
  • Vapor pressure:
    0,07 hPa (20°C)
  • Solubility:
    40 g/l
  • Ignition temperature:
    435 °C
  • Storage temperature:
    2 - 25 °C
  • Molar mass:
    108,14 g/mol

Specification Test Results

Related Information

Frequently Bought Together