Chemical resistant gloves, Showa 3415
Supplier: Showa
Certificates
About this item
These seamless knitted gloves with flexible, consistent neoprene coating provide protection against a wide range of chemicals, including acids, caustics, solvents, grease and oil.
- Fully coated
- Rough particle grip
- Gauntlet cuff for protection of the forearm
Recommended industries: Mechanical, automotive, petrochemical.
Certifications: EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5; Cat. III
Specifications
- Catalog No:
- 112-5781
- 112-5783
- 112-5782
- 112-5780
- Model:
- 3415
- 3415
- 3415
- 3415
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Rough particle grip
- Rough particle grip
- Rough particle grip
- Rough particle grip
- Gauge:
- 15
- 15
- 15
- 15
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5
- EN 388:2016 (3031X), EN ISO 374-1 Type A (AJKLMT), EN ISO 374-5