Safety spectacles, Solis
Catalog # 111-0156
Supplier: Bollé Safety
Specifications
- Type:Anti-UV (5-3,2), anti-scratch, polarised coating
- Colour:Black
- Environmentally Preferable:
- Lenses:Smoke PC
- Pk:1
Specifications
About this item
Solis spectacles offer a wrap-around style with the benefits of development of modern materials.
- Non slip TIPGRIP temples
- Non slip bridge
- Integral side shields
- Polarised coating
Weight: 25 g
Certifications: EN 166, EN 172
Delivery: Supplied with microfibre pouch.