Order Entry
Denmark
ContactUsLinkComponent
Neutralizing agent, RBS A 375
Neutralizing agent, RBS A 375
Catalog # CHEPRBSA375/E123
Supplier:  CHEMICAL
undefined
Neutralizing agent, RBS A 375
Catalog # CHEPRBSA375/E123
Supplier:  CHEMICAL
Specifications
  • Description:
    Neutralizing agent, RBS A 375
  • Environmentally Preferable:
  • Pk:
    5 L
  • pH range:
    2.6 (0.5%)
Specifications
About this item
Improves the cleaning results in washing machines - labwashers.
  • Concentrated solution
  • Surfactant-free
  • Based on citric acid
RBS A 375 solution allows neutralisation and removal of residual alkaline traces and mineral salts from the cleaned items and the internal components of the washing machine. Can also be used in the pre-wash cycle in case of heavy residues. Improves the rinsing cycle and brightens the cleaned items.
Ordering information: Available in box of 4 × 5 litres.