Order Entry
Denmark
ContactUsLinkComponent
Alkaline detergent, RBS T 105
Alkaline detergent, RBS T 105
Catalog # CHEPT105/E123
Supplier:  CHEMICAL
undefined
Alkaline detergent, RBS T 105
Catalog # CHEPT105/E123
Supplier:  CHEMICAL

Specifications

  • Description:
    Alkaline detergent, RBS T 105
  • Environmentally Preferable:
  • Pk:
    5 L
  • pH range:
    11.8 (2%)

Specifications

About this item

Suitable for efficient manual and ultrasonic cleaning of glassware and laboratory equipement made of stainless steel, plastic and porcelain.

  • High detergency
  • Low use concentration

RBS T 105 is recommended for manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment made of glass, plastic, stainless steel and porcelain. It efficiently removes organic residues, blood and biological residues, fats, oils, distillation residues etc. It rinses easily without leaving any interfering residues.

Ordering information: Available in box of 4 × 5 litres. Please contact Avantor for details.
Caution: Not recommended for use on white metals.